PIG® Traffic Mat® absorbent rugs
Supplier: NEW PIG
Certificates
About this item
Keep floors dry, even in areas with forklift traffic.
- 100% polypropylene material is heat fused and needle punched for incredible durability for walk-on and wheeled traffic
- Our most durable mat won't rip, shred or fray, even under heavy forklift traffic!
- Poly backing prevents absorbed liquids from reaching floors or counters
- Absorbs and retains oils, coolants, solvents and water
- Dark grey colour hides grime, so mat stays on the job longer
Rolls are ideal for walkways and large areas to soak up leaks and overspray. Cuts easily with scissors or utility knife. Can be incinerated after use to reduce waste or for fuels blending.