HPLC columns, NUCLEOSIL® C8 HD
NUCLEOSIL® C8 HD is an octyl modified silica phase with a high density coverage, using for HPLC.
- Robust spherical silica gel
- Hydrophobic, weakly polar, endcapped phase for RP chromatography
- Separation mechanism based on hydrophobic interactions
EC guard columns are used for analytical EC columns with the column protection system guard column holder (554-5851).
- Catalog No:
- MANA721602.40
- MANA720196.46
- MANA720196.40
- MANA720194.46
- MANA761100.40
- MANA721500.40
- MANA721071.30
- Kohlenstoffbelastung:
- 13%
- 13%
- 13%
- 13%
- 13%
- 13%
- 13%
- Endcapped:
- Ja, Ja
- Ja, Ja
- Ja, Ja
- Ja, Ja
- Ja, Ja
- Ja, Ja
- Ja, Ja
- pH-Bereich:
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- USP-Code:
- L7, L7
- L7, L7
- L7, L7
- L7, L7
- L7, L7
- L7, L7
- L7, L7
Frequently Bought Together