Reversed phase HPLC columns, biochemical analysis, NUCLEOSIL® MPN
Silica based reversed phase materials with monomerically bonded alkyl chains for RP chromatography that is ideal for seperation of biological macromolecules.
- Based on robust spherical silica gel
- Predominantly hydrophobic forces with a small portion of hydrophilic interactions
- Eluent in column methanol
NUCLEOSIL® C18 MPN columns with particle size of 100 Å are suited for small hydrophobic peptides; C4 MPN with 300 Å for larger hydrophobic peptides and very different proteins.
- Catalog No:
- MANA720231.40
- MANA720245.40
- Description:
- C18 MPN
- C4 MPN
- Max. pressure:
- 250 bar
- 250 bar
- pH range:
- 2 – 8
- 2 – 8
- USP code:
- L1
- L26