Capillary blood collection tubes, MiniCollect®, Vacuette®
Supplier: Greiner Bio-One
Certificates
About this item
PP, translucent, with cap.
- With or without additives
MiniCollect® tubes are ideal for collecting, transporting and processing capillary blood. Capillary blood collection is especially recommended in patients with fragile veins, young children, geriatric patients and in situations where only a small blood sample is required. MiniCollect® tubes are available with a volume of 0,25 to 1 ml and with a range of additives.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904