Erythrocyte sedimentation system, Sedi-Rate
Supplier: KARTELL
Certificates
About this item
Sedi-Rate allows a rapid and safe execution of the ESR test.
- PP tube (lenght 86 mm) with label complete with pink fiber cap containing sodium citrate
- Pipette in PS is silk-screened with a graduation scale
- Piston allowing a quick filling of the pipette thanks to the pressure within the tube
According to the Westergren method.