Order Entry
Germany
ContactUsLinkComponent
pH/mV/°C/ion meter, handheld, 3205
pH/mV/°C/ion meter, handheld, 3205
Catalog # 662-0070
Supplier:  Cole-Parmer
pH/mV/°C/ion meter, handheld, 3205
Catalog # 662-0070
Supplier:  Cole-Parmer

Specifications

  • Temperature range:
    −10...+105 °C
  • Description:
    pH/ion meter, handheld, 3205
  • Model:
    3205
  • Temperature accuracy:
    ±0,5 °C
  • Temperature resolution:
    0,1 °C
  • Automatic buffer recognition:
    4, 7 and 10,05 (with manual override)
  • Connections:
    S7, BNC
  • ISE calibration:
    Multipoint calibration 1 to 5 points
  • ISE units:
    %, ppm, mg/l, moles, none
  • ISE range:
    1×10⁻⁹ - 9,99×10⁹ ppm
  • Power supply:
    3 AA batteries
  • Temperature compensation:
    Automatic (0 - 100 °C)
  • Weight:
    0,37 kg
  • W×D×H:
    80×60×200 mm
  • mV accuracy:
    ±1
  • mV range:
    ±1999
  • mV resolution:
    1
  • pH accuracy:
    ±0,02
  • pH calibration:
    User selectable, 1 or 2 point
  • pH range:
    −2,00...+16,00
  • pH resolution:
    0,01
  • Pk:
    1

Specifications

About this item

The 3205 portable ion meter is dedicated to the rapid, easy and reliable determinations of ammonium, calcium, chloride, fluoride and nitrate ions with concentrations displayed in mg/l, ppm, %, moles, activity or no units.

  • Combined pH and ion portable, waterproof meter
  • Up to 2 point pH and 5 point ion calibration
  • Automatic buffer recognition
  • Fully GLP compliant
  • RS 232 interface using optional cradle
  • Six language options

When performing dedicated ion analysis appropriate ATC parameters will be automatically selected and when combined with the very stable electrode readings all calibrations can be performed in the laboratory prior to analysis in the field. Although customised for dedicated ion analysis the 3205 can also perform measurements of pH, mV and other, non-dedicated ion measurements.

Delivery: Supplied with pH/temperature probe (027 227), buffer sachets, waterproof connector to S7 electrode lead (013 161), waterproof connector to BNC electrode lead and batteries.