Orders LinkContactUsLinkComponent
Cell scrapers
Cell scrapers
Cell scraper with two-position blade, 250 mm
Catalog # SARS83.3951
Marketsource
Supplier:   SARSTEDT
Cell scrapers
Catalog # SARS83.3951
Supplier:  SARSTEDT
Marketsource
Cell scrapers
Cell scraper with two-position blade, 250 mm
Price
132,00 €
Log in for availability
1 * 100 ST
Product Details & Documents
Cell scraper with soft, flexible blade for gentle removal of monolayer cultures, minimising cell damage.
  • Easy and complete separation of monolayer due to extra thin and flexible blade
  • When downward pressure is applied to the handle the blade can remove all cells in its path in one pass
  • Sterilised by gamma irradiation, individually wrapped
Specifications
  • Description:
    Cell scraper with two-position blade
  • Seq:
    01
  • Total length:
    250 mm