Cleanroom anti-fatigue mats, Ergomat Nitro
Supplier: ERGOMAT
Certificates
About this item
Ergomat® Nitro with anti-slip cone profile is made from pure nitrile rubber and is designed to increase comfort while reducing trip hazards, body pain, quality issues, and sick leave. It is ideal for usage in the electronics industry or for areas where static control is crucial due to sensitive equipment.
- Upper side with cones
- Autoclavable, withstands 600 °C (welding)
- Chemical exposure will not affect the state of the mat; no curling up, swelling, or expanding
- Moulded bevelled edges prevent curling, eliminates trip hazards
- Raw virgin material guarantee durability and ergonomic benefits
- Free from silicone and latex
Suitable for use in Class ISO 5 (100) cleanrooms.
Colour: Black
Certifications: Meets EN ISO/USDA/CFIA standards.