Order Entry
Germany
Orders LinkContactUsLinkComponent
hearing--hero.jpg
Avantor® Presents 3M Solutions: From Detection to Protection & Validation
Hearing protection
Easy solutions to complex hearing protection challenges
3M Solutions - Hearing Protection | Avantor
As a key strategic partner to Avantor® in personal protective equipment and safety solutions, 3M is passionate about protecting the health and safety of workers in all types of industries and workforce environments.
3M, as a global authority in helping you solve your most complex hearing protection challenges, has launched great tools and services to assist you — from detection to protection to validation.
Use this new platform to download these new tools and services to complement Avantor's offering of personal protection equipment.
 Detection
Detecting noise levels in the workplace is the first step to ensuring your workforce is protected.
Complying with regulations, selecting appropriate protection and analyzing noise control options are all easier with 3M solutions for exposure assessment, noise analysis and employee engagement.
3M's accurate, reliable and flexible instrumentation coupled with powerful software is the innovative solution for your noise detection challenge.
Protection and Validation with Hearing Protection 540.png
Detecting noise levels in the workplace is the first step to ensuring your workforce is protected
Complying with regulations, selecting appropriate protection and analyzing noise control options are all easier with 3M solutions for exposure assessment, noise analysis and employee engagement. 3M's accurate, reliable and flexible instrumentation coupled with powerful software is the innovative solution for your noise detection challenge.
hearing.png
3M E-A-R Classic Earplugs are disposable and designed for insertion into the ear canal to help reduce exposure to hazardous levels of noise and loud sound up to 31 dB SNR. These products are available in corded and uncorded versions.
Hearing2.png
3M 1100 Earplugs are disposable foam earplugs designed for insertion into the ear canal to help reduce exposure to hazardous levels of noise up to 35 dB, tested to EN 352-2:2020.
Workers.png
3M PELTOR WS ALERT X Headsets are Bluetooth® headsets with a noise-cancelling boom microphone that connects to your mobile phone for hands-free calls and streaming in noisy environments. These headsets have a level-dependent function for ambient listening, and they sync to a mobile app for easy setup and adjustment.
3M™ PELTOR™ WS™ LiteCom Pro III Headsets.png
3M PELTOR WS LiteCom Pro III Headsets are our high-performance hearing protectors with a built-in analog and digital programmable two-way communication radio, level-dependent functionality for ambient listening, Bluetooth® multipoint connectivity, a lithium ion rechargeable battery and an external connection jack. Available in headband, neckband and helmet-mounted models.
BUY PRODUCTS