Order Entry
Germany
Orders LinkContactUsLinkComponent
Laminar-Air-Flow-Systems.png
Laminar Air Flow Systems
Laminar flow cabinets are carefully designed to protect very sensitive products from external contamination or the user from contamination by the product - or both. This very clean working atmosphere is obtained by blowing air through HEPA filters. The airstream can be vertical or horizontal, depending on the application.
Laminar Air Flow Systems | Avantor
Product selection