ContactUsLinkComponent
Phenols Mixture - 19 components: 100mg/l each of 2-Chlorophenol [CAS:95-57-8] 3-Chlorophenol [CAS:108-43-0] 4-Chlorophenol [CAS:106-48-9] 2,3-Dichlorophenol [CAS:576-24-9] 2,4-Dichlorophenol [CAS:120-83-2] 2,5-Dichlorophenol [CAS:583-78-8] 1 * 1 mL
Catalog # ACSDF224433
Supplier:  CPACHEM
New Product
Phenols Mixture - 19 components: 100mg/l each of 2-Chlorophenol [CAS:95-57-8] 3-Chlorophenol [CAS:108-43-0] 4-Chlorophenol [CAS:106-48-9] 2,3-Dichlorophenol [CAS:576-24-9] 2,4-Dichlorophenol [CAS:120-83-2] 2,5-Dichlorophenol [CAS:583-78-8] 1 * 1 mL
Catalog # ACSDF224433
Supplier:  CPACHEM