ContactUsLinkComponent
RadEye B20-ER, Alpha, Beta, Gamma Surveymeter, extended measuring range up to 500 kcps, Beta and Gamma dose rate up to 100 mSv/h 10 Rem/h, including hard shell transport case 1 * 1 items
Catalog # HERA425068510
Supplier:  Thermo Scientific
New Product
RadEye B20-ER, Alpha, Beta, Gamma Surveymeter, extended measuring range up to 500 kcps, Beta and Gamma dose rate up to 100 mSv/h 10 Rem/h, including hard shell transport case 1 * 1 items
Catalog # HERA425068510
Supplier:  Thermo Scientific