ContactUsLinkComponent
arget Protein,,Granzyme H Clonality,,Polyclonal Storage/Stability,,-20°C/1 year Application,,WB,IHC,IF/ICC,ELISA Recommended Dilution,,WB 1:1000 IHC 1:200, IF/ICC 1:100-1:500 Concentration,,1 mg/ml Reactive Species,,Human,Mouse Host Species,,Rabbit I 1 * 200 µG
Catalog # GENOITA2121-200U-F
Supplier:  G-Biosciences
New Product
arget Protein,,Granzyme H Clonality,,Polyclonal Storage/Stability,,-20°C/1 year Application,,WB,IHC,IF/ICC,ELISA Recommended Dilution,,WB 1:1000 IHC 1:200, IF/ICC 1:100-1:500 Concentration,,1 mg/ml Reactive Species,,Human,Mouse Host Species,,Rabbit I 1 * 200 µG
Catalog # GENOITA2121-200U-F
Supplier:  G-Biosciences