ContactUsLinkComponent
Target Protein,,Granzyme H Clonality,,Polyclonal Storage/Stability,,-20°C/1 year Application,,WB,IHC,IF/ICC,ELISA Recommended Dilution,,WB 1:1000 IHC 1:200, IF/ICC 1:100-1:500 Concentration,,1 mg/ml Reactive Species,,Human,Mouse Host Species,,Rabbit 1 * 100 µG
  GENOITA2121-100U-F
New Product
Target Protein,,Granzyme H Clonality,,Polyclonal Storage/Stability,,-20°C/1 year Application,,WB,IHC,IF/ICC,ELISA Recommended Dilution,,WB 1:1000 IHC 1:200, IF/ICC 1:100-1:500 Concentration,,1 mg/ml Reactive Species,,Human,Mouse Host Species,,Rabbit 1 * 100 µG
  GENOITA2121-100U-F