Bacillus spizizenii WDCM 00003 Vitroids 80-130 CFU mean value range, certified reference material, suitable for microbiology, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland 1 * 10 items
Catalog # VT000033-10EA
Supplier: Merck