ContactUsLinkComponent
HiTrap™ Protein Select™ column 5 × 5 mL. Protein Select™ is an affinity chromatography resin for purifying recombinant proteins using the self-cleaving Cytiva™ Protein Select™ tag 1 * 25 mL
Catalog # 17-5421-54
Supplier:  Cytiva
New Product
HiTrap™ Protein Select™ column 5 × 5 mL. Protein Select™ is an affinity chromatography resin for purifying recombinant proteins using the self-cleaving Cytiva™ Protein Select™ tag 1 * 25 mL
Catalog # 17-5421-54
Supplier:  Cytiva