ContactUsLinkComponent
Enclosed form, -25+100:0.2°C, with special inner capillary coating, with non-wetting blue special filling, 450x9-9.5mm, fully immersible, Richter closure with button, calibrated with DKD certificate (DAkkS) with 2 test points 1 * 1 items
Catalog # AMARL25950CBDKD2CL
Supplier:  Amarell
New Product
Enclosed form, -25+100:0.2°C, with special inner capillary coating, with non-wetting blue special filling, 450x9-9.5mm, fully immersible, Richter closure with button, calibrated with DKD certificate (DAkkS) with 2 test points 1 * 1 items
Catalog # AMARL25950CBDKD2CL
Supplier:  Amarell