ContactUsLinkComponent
6 components: Fluorides (F-) 75mg/l Chlorides (Cl-) 1250mg/l Phosphates (PO43-) 10mg/l Sulphates (SO42-) 1500mg/l Nitrates (NO3-) 150mg/l Bromides (Br-) 6mg/l in Water CRM, ISO 17034 and ISO 17025 Conservation 12 months. Store under normal 1 * 500 mL
Catalog # CMPLRM122789L5
New Product
6 components: Fluorides (F-) 75mg/l Chlorides (Cl-) 1250mg/l Phosphates (PO43-) 10mg/l Sulphates (SO42-) 1500mg/l Nitrates (NO3-) 150mg/l Bromides (Br-) 6mg/l in Water CRM, ISO 17034 and ISO 17025 Conservation 12 months. Store under normal 1 * 500 mL
Catalog # CMPLRM122789L5