Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
STANDARD PLATE COUNT AGAR A.P.H.A 5KG 1 * 5 kg
OXOICM0463T
:
THERMO FISHER DIAGNOSTICS
New Product
STANDARD PLATE COUNT AGAR A.P.H.A 5KG 1 * 5 kg
OXOICM0463T
:
THERMO FISHER DIAGNOSTICS
New Product
Frequently Bought Together
ToxinEraserTM Endotoxin Removal Resin 1 * 1 mL
: Genscript
Culture media, ready to use, Compact Dry
: Shimadzu Diagnostics Europe
VWR®, Lockable contact plate for pharmaceutical industries
: VWR Chemicals
LUGOL PVP SOLUTION-250 ML 1 * 1 items
: LIOFILCHEM