ContactUsLinkComponent
Pur-A-Lyzer Maxi Dialysis Kit Maxi 12000, capacity 0.1-3 mL, pkg of 50 Maxi dialysis tubes, MWCO 12-14 kDa 1 * 1 KIT
Catalog # SIALPURX12050-1KT
Supplier:  Merck
New Product
Pur-A-Lyzer Maxi Dialysis Kit Maxi 12000, capacity 0.1-3 mL, pkg of 50 Maxi dialysis tubes, MWCO 12-14 kDa 1 * 1 KIT
Catalog # SIALPURX12050-1KT
Supplier:  Merck