Cesium chloride-aluminium nitrate buffer solution 50 g/l CsCl / 250 g/l Al(NO3)3.9H2O ,,,, ,, Conservation 24 months.,,,, Store under normal laboratory conditions, at temperatures between 15°C to 25°C 1 * 1 L
Catalog # CMPLIB637.1L
Supplier: CUSTOM MADE CHEMICALS LAB