ContactUsLinkComponent
Quality Control Standard of Mineral Oils (ISO 9377-2-Mod) 2 components: 5000mg/l each of Mineral Oil [CAS:74869-22-0] Diesel Oil [CAS:68334-30-5] in n-Hexane ,,,, CRM, ISO 17034 and ISO 17025,,,, Conservation 24 months.,,,,,, Store in a refrigerato 1 * 5 mL
Catalog # ACSDF109661
Supplier:  CPACHEM
New Product
Quality Control Standard of Mineral Oils (ISO 9377-2-Mod) 2 components: 5000mg/l each of Mineral Oil [CAS:74869-22-0] Diesel Oil [CAS:68334-30-5] in n-Hexane ,,,, CRM, ISO 17034 and ISO 17025,,,, Conservation 24 months.,,,,,, Store in a refrigerato 1 * 5 mL
Catalog # ACSDF109661
Supplier:  CPACHEM