ContactUsLinkComponent
1/4 GC ferrule 6.0 mm ID 85% Vespel / 15% Graphite PK10 1 * 10 items
Catalog # SGEA215460
New Product
1/4 GC ferrule 6.0 mm ID 85% Vespel / 15% Graphite PK10 1 * 10 items
Catalog # SGEA215460