ContactUsLinkComponent
1/16 GC ferrule 0.3 and 0.5 mm ID (long) 85% Vespel / 15% Graphite PK10 1 * 10 items
Catalog # SGEA215135
New Product
1/16 GC ferrule 0.3 and 0.5 mm ID (long) 85% Vespel / 15% Graphite PK10 1 * 10 items
Catalog # SGEA215135