ContactUsLinkComponent
PEEK TUBE BLACK 1/16 OD X 0.040 ID, LENGTH 500 METERS 1 * 1 items
Catalog # SGEA1302040006500M
New Product
PEEK TUBE BLACK 1/16 OD X 0.040 ID, LENGTH 500 METERS 1 * 1 items
Catalog # SGEA1302040006500M