ContactUsLinkComponent
,,13 components: 100mg/l each of Al B Ba Be Ca Cr Fe K Mg Mn Na Ti Li in HNO3 2% ,, ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 24 months.,, ,,Ambient Temp.,, 1 * 100 mL
Catalog # CMPLRM122046L1
New Product
,,13 components: 100mg/l each of Al B Ba Be Ca Cr Fe K Mg Mn Na Ti Li in HNO3 2% ,, ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 24 months.,, ,,Ambient Temp.,, 1 * 100 mL
Catalog # CMPLRM122046L1