ContactUsLinkComponent
Muffle Furnace LT15/13/C550 Tmax1300 with lift door and over-temperature limiter and thermocouple port in the rear wall 1 * 1 items
Catalog # 455-3300
Supplier:  NABERTHERM
New Product
Muffle Furnace LT15/13/C550 Tmax1300 with lift door and over-temperature limiter and thermocouple port in the rear wall 1 * 1 items
Catalog # 455-3300
Supplier:  NABERTHERM