ContactUsLinkComponent
Muffle Furnace LT15/13/C550 Tmax1300 with lift door and protective gas connection, over-temperature limiter and thermocouple port in the rear wall 1 * 1 items
Catalog # 455-3295
Supplier:  NABERTHERM
New Product
Muffle Furnace LT15/13/C550 Tmax1300 with lift door and protective gas connection, over-temperature limiter and thermocouple port in the rear wall 1 * 1 items
Catalog # 455-3295
Supplier:  NABERTHERM