ContactUsLinkComponent
Muffle Furnace L5/13/B510 Tmax1300 with flap door and protective gas connection and over-temperature limiter 1 * 1 items
Catalog # 455-3105
Supplier:  NABERTHERM
New Product
Muffle Furnace L5/13/B510 Tmax1300 with flap door and protective gas connection and over-temperature limiter 1 * 1 items
Catalog # 455-3105
Supplier:  NABERTHERM