ContactUsLinkComponent
Muffle Furnace L40/12/B510 Tmax1200 with flap door and protective gas connection and over-temperature limiter 1 * 1 items
Catalog # 455-3053
Supplier:  NABERTHERM
New Product
Muffle Furnace L40/12/B510 Tmax1200 with flap door and protective gas connection and over-temperature limiter 1 * 1 items
Catalog # 455-3053
Supplier:  NABERTHERM