ContactUsLinkComponent
Muffle Furnace L15/11/B510 Tmax1100 with flap door and protective gas connection, over-temperature limiter and thermocouple port in the rear wall 1 * 1 items
  455-2814
New Product
Muffle Furnace L15/11/B510 Tmax1100 with flap door and protective gas connection, over-temperature limiter and thermocouple port in the rear wall 1 * 1 items
  455-2814