Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
GFP ELISA Kit

GFP ELISA Kit

GFP ELISA Kit, 10×96 tests
Katalogové číslo ABCAAB171581-10X96
New Product
Dodavatel:   Abcam
GFP ELISA Kit
Katalogové číslo ABCAAB171581-10X96
Dodavatel:  Abcam
New Product
Číslo CAS:  
GFP ELISA Kit
GFP ELISA Kit

GFP ELISA Kit, 10×96 tests

Přejít na:
Číslo CAS:  
Price
180 600,00 Kč
Přihlásit se pro dostupnost
1 * 10 Tests
Product Details & Documents

GFP ELISA kit is a single-wash 90-min SimpleStep ELISA® used to quantify Green Fluorescence protein with a sensitivity of 2.8 pg/ml. The assay uses a simple Mix-Wash-Read protocol with just one incubation and wash step. Colorimetric assay - 450 nm readout; works on any plate reader. Different formats for different needs: 10×96-well plates for bulk orders and 384-well for higher throughput. Broad sample compatibility, works with tissue lysate and cell lysate.

Documents

Specifikace

Specifikace

  • Description:
    GFP ELISA Kit
  • Tests per kit:
    10×96 tests
  • Primary antibody reactivity:
    Species independent
  • Sample type:
    Tissue Lysate, Cell Lysate
  • Detection range:
    18,8 - 1200 pg/ml
  • Storage temperature:
    4 °C
  • Assay Type:
    Sandwich
  • Time to Results:
    1 hr 30 min
  • Cat. No.:
    ABCAAB171581-10X96
  • Assay duration:
    One step
  • Detection method:
    Colorimetric
  • Sensitivity:
    2,8 pg/ml
  • Format:
    Pre-coated
  • Target protein:
    GFP
  • Regulatory status:
    RUO

Specifikace