ContactUsLinkComponent
Pump module VDL 23 with VP5 EX vacuum pump - 230 V / 50-60 Hz version, pump cabinet for vacuum pumps. Includes ATEX chemical diaphragm pump with a nominal pumping speed of 1,9 m /h, final pressure of 12 mbar, 230 V/50 Hz, and connnection kit with sta 1 * 1 items
Catalog # BIND8012-2628
Supplier:  Binder
New Product
Pump module VDL 23 with VP5 EX vacuum pump - 230 V / 50-60 Hz version, pump cabinet for vacuum pumps. Includes ATEX chemical diaphragm pump with a nominal pumping speed of 1,9 m /h, final pressure of 12 mbar, 230 V/50 Hz, and connnection kit with sta 1 * 1 items
Catalog # BIND8012-2628
Supplier:  Binder