ContactUsLinkComponent
Pump module VD 23 with vacuum pump VP2 - 230 V / 50-60 Hz version, pump cabinet for vacuum pumps, with switchable socket. Includes chemical diaphragm pump with nominal pumping speed of 3,4 m /h, final pressure of 1,5 mbar, and connection kit VD VP1-V 1 * 1 items
Catalog # BIND8012-2617
Supplier:  Binder
New Product
Pump module VD 23 with vacuum pump VP2 - 230 V / 50-60 Hz version, pump cabinet for vacuum pumps, with switchable socket. Includes chemical diaphragm pump with nominal pumping speed of 3,4 m /h, final pressure of 1,5 mbar, and connection kit VD VP1-V 1 * 1 items
Catalog # BIND8012-2617
Supplier:  Binder