ContactUsLinkComponent
VDL VP5 PTFE connection kit - for VP5 EX vacuum pump. Include a 1,9 m long antistatic PTFE hose with a DN 16 small flange on both sides, two clamping rings, and two universal centering rings. 1 * 1 KIT
Catalog # BIND8012-2614
Supplier:  Binder
New Product
VDL VP5 PTFE connection kit - for VP5 EX vacuum pump. Include a 1,9 m long antistatic PTFE hose with a DN 16 small flange on both sides, two clamping rings, and two universal centering rings. 1 * 1 KIT
Catalog # BIND8012-2614
Supplier:  Binder