ContactUsLinkComponent
Connection kit VDL VP5 stainless steel - for VP5 EX vacuum pump. Includes 1,9 m long stainless steel hose with a DN 16 small flange on both sides, two clamping rings, and two universal centering rings. 1 * 1 KIT
Catalog # BIND8012-2724
Supplier:  Binder
New Product
Connection kit VDL VP5 stainless steel - for VP5 EX vacuum pump. Includes 1,9 m long stainless steel hose with a DN 16 small flange on both sides, two clamping rings, and two universal centering rings. 1 * 1 KIT
Catalog # BIND8012-2724
Supplier:  Binder