ContactUsLinkComponent
Vacuum pump VP5 EX - , ATEX chemical diaphragm pump with nominal pumping speed 1,9 m /h, final pressure 12 mbar, 230V/50Hz. ATEX conformity: pump chamber (pumped gases): II 2G Ex h IIC T3 Gb X External space with inert gas purge: II 2G Ex h IIB T4 G 1 * 1 items
Catalog # BIND5013-0258
Supplier:  Binder
New Product
Vacuum pump VP5 EX - , ATEX chemical diaphragm pump with nominal pumping speed 1,9 m /h, final pressure 12 mbar, 230V/50Hz. ATEX conformity: pump chamber (pumped gases): II 2G Ex h IIC T3 Gb X External space with inert gas purge: II 2G Ex h IIB T4 G 1 * 1 items
Catalog # BIND5013-0258
Supplier:  Binder