ContactUsLinkComponent
Vacuum pump VP2 - chemical diaphragm pump with nominal pumping speed of 3,4 m /h and a final pressure of 1,5 mbar. Switchable between 230V/50Hz and 120V/60Hz. 1 * 1 items
Catalog # BIND5013-0257
Supplier:  Binder
New Product
Vacuum pump VP2 - chemical diaphragm pump with nominal pumping speed of 3,4 m /h and a final pressure of 1,5 mbar. Switchable between 230V/50Hz and 120V/60Hz. 1 * 1 items
Catalog # BIND5013-0257
Supplier:  Binder