ContactUsLinkComponent
Vacuum pump VP1 - chemical diaphragm pump with nominal pumping speed of 2,0 m /h and a final pressure of 7 mbar. Switchable between 230V/50Hz and 120V/60Hz. 1 * 1 items
Catalog # BIND5013-0256
Supplier:  Binder
New Product
Vacuum pump VP1 - chemical diaphragm pump with nominal pumping speed of 2,0 m /h and a final pressure of 7 mbar. Switchable between 230V/50Hz and 120V/60Hz. 1 * 1 items
Catalog # BIND5013-0256
Supplier:  Binder