Water CHROMASOLV, for HPLC 6 x 1L 1 * 6 items
Katalogové číslo HONC39253-6X1L
Supplier: Honeywell Chemicals
New Product
Water CHROMASOLV, for HPLC 6 x 1L 1 * 6 items
Katalogové číslo HONC39253-6X1L
Supplier: Honeywell Chemicals
New Product
Frequently Bought Together
Assistent Brushes with Cotton Tip Bristles
Supplier: GLASWARENFABRIK KARL HECHT
Single-use syringes, 2-piece, HENKE-JECT®
Supplier: HENKE SASS WOLF
Cleanroom wipes, TechniCloth®
Supplier: Texwipe
Ethanol, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Ethanol absolute ≥99.5% USP, Ph. Eur.
Supplier: VWR Chemicals
Ethanol 96% vol Ph. Eur.
Supplier: VWR Chemicals
Pipette tips, Finntip™
Supplier: Thermo Fisher Scientific
Nitric acid 0.2 mol/l (0.2 N)
Supplier: VWR Collection
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals