ContactUsLinkComponent
Extremely stable, long overhanging stand for observation of particularly large specimens to be matched with a focusing system and stereozoom head. Smooth horizontal and vertical ovements Horizontal arm: 415 mm Maximum sample height: 400 mm Swivelling 1 * 1 items
Catalog # OPTKSZ-STL1
Supplier:  OPTIKA
New Product
Extremely stable, long overhanging stand for observation of particularly large specimens to be matched with a focusing system and stereozoom head. Smooth horizontal and vertical ovements Horizontal arm: 415 mm Maximum sample height: 400 mm Swivelling 1 * 1 items
Catalog # OPTKSZ-STL1
Supplier:  OPTIKA