ContactUsLinkComponent
26 components: 100ug/ml each of Al As B Ba Bi Ca Cd Co Cr Cu Fe K Li Mg Mn Mo Na Ni Pb Sb Sn Sr Ti V Zn Zr in HNO3 10% HF 1% CRM, ISO 17034 and ISO 17025 1 * 100 mL
Katalogové číslo CMPLRM118885L1
New Product
26 components: 100ug/ml each of Al As B Ba Bi Ca Cd Co Cr Cu Fe K Li Mg Mn Mo Na Ni Pb Sb Sn Sr Ti V Zn Zr in HNO3 10% HF 1% CRM, ISO 17034 and ISO 17025 1 * 100 mL
Katalogové číslo CMPLRM118885L1