ContactUsLinkComponent
Scientific Line, Underbench ATEX Freezer Solid door, 3 Freezer Drawers, temperature stability:+/-5°C, temperature range: -9/-30 °C, gross/net volume (L): 101/71, dimensions in mm (W/D/H): 597 / 605 / 819 1 * 1 items
Catalog # LIEBSFTFG1501_740
Supplier:  Liebherr
New Product
Scientific Line, Underbench ATEX Freezer Solid door, 3 Freezer Drawers, temperature stability:+/-5°C, temperature range: -9/-30 °C, gross/net volume (L): 101/71, dimensions in mm (W/D/H): 597 / 605 / 819 1 * 1 items
Catalog # LIEBSFTFG1501_740
Supplier:  Liebherr