ContactUsLinkComponent
Mouse Collagen II ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determin ation of mouse Collagen II in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA270880-48
Supplier:  Antibodies.com
New Product
Mouse Collagen II ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determin ation of mouse Collagen II in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA270880-48
Supplier:  Antibodies.com