ContactUsLinkComponent
Rat BDNF ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat BDNF in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological fluids. 1 * 48 Tests
Katalogové číslo ANTIA1656-48
Dodavatel:  Antibodies.com
New Product
Rat BDNF ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat BDNF in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological fluids. 1 * 48 Tests
Katalogové číslo ANTIA1656-48
Dodavatel:  Antibodies.com