ContactUsLinkComponent
Human ADA ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human ADA in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
  ANTIA2458-48
New Product
Human ADA ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human ADA in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
  ANTIA2458-48