ContactUsLinkComponent
Llama Interferon gamma Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detect ion antibody, Streptavidin-AP, recombinant llama Interferon gamma ELISA standard, and standard reconstitution buffer. 1 * 576 Tests
Llama Interferon gamma Matched Antibody Pair Kit
Catalog # ANTIA270422-696
Supplier:  Antibodies.com
New Product
CAS Number:  
Llama Interferon gamma Matched Antibody Pair Kit
Catalog # ANTIA270422-696
Supplier:  Antibodies.com
New Product
CAS Number:  
About this item
Frequently Bought Together