Rat Amyloid beta Peptide 1-40 ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quanti tative determination of rat Amyloid beta Peptide 1-40 in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA1804-48
Supplier: Antibodies.com