ContactUsLinkComponent
Human alpha 1 Acid Glycoprotein/AGP ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qua ntitative determination of human alpha 1 Acid Glycoprotein/AGP in serum, plasma, tissue homogenates, cell lysates, ce 1 * 96 Tests
Catalog # ANTIA1970-96
Dodavatel:  ANTIBODIES.COM
New Product
Human alpha 1 Acid Glycoprotein/AGP ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro qua ntitative determination of human alpha 1 Acid Glycoprotein/AGP in serum, plasma, tissue homogenates, cell lysates, ce 1 * 96 Tests
Catalog # ANTIA1970-96
Dodavatel:  ANTIBODIES.COM