ContactUsLinkComponent
Bovine Transferrin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determi nation of bovine Transferrin in serum, plasma or other biological fluids. 1 * 48 Tests
  ANTIA270961-48
New Product
Bovine Transferrin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determi nation of bovine Transferrin in serum, plasma or other biological fluids. 1 * 48 Tests
  ANTIA270961-48